Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proprotein Convertase 9/PCSK9 Antibody (106), DyLight 650, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290497C
Description
Proprotein Convertase 9/PCSK9 Monoclonal specifically detects Proprotein Convertase 9/PCSK9 in Mouse samples. It is validated for ELISA.Specifications
Proprotein Convertase 9/PCSK9 | |
Monoclonal | |
DyLight 650 | |
EC 3.4.21, EC 3.4.21.111, FH3neural apoptosis regulated convertase 1, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Proprotein Convertase 9/PCSK9 (NP_705793.1; Met 1-Gln 694). | |
0.1 mL | |
Cholesterol Metabolism, Lipid and Metabolism | |
255738 | |
Mouse | |
Purified |
ELISA | |
106 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human PCSK9 Rhesus PCSK9 Human PCSK1 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction