Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propyl gallate 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
121-79-9
C10H12O5
212.2
MFCD00002196
ZTHYODDOHIVTJV-UHFFFAOYSA-N
propyl gallate,n-propyl gallate,progallin p,tenox pg,nipagallin p,gallic acid, propyl ester,gallic acid propyl ester,nipa 49,benzoic acid, 3,4,5-trihydroxy-, propyl ester,3,4,5-trihydroxybenzene-1-propylcarboxylate
4947
CHEBI:10607
propyl 3,4,5-trihydroxybenzoate
CCCOC(=O)C1=CC(=C(C(=C1)O)O)O
121-79-9
C10H12O5
212.2
MFCD00002196
ZTHYODDOHIVTJV-UHFFFAOYSA-N
propyl gallate,n-propyl gallate,progallin p,tenox pg,nipagallin p,gallic acid, propyl ester,gallic acid propyl ester,nipa 49,benzoic acid, 3,4,5-trihydroxy-, propyl ester,3,4,5-trihydroxybenzene-1-propylcarboxylate
4947
CHEBI:10607
propyl 3,4,5-trihydroxybenzoate
CCCOC(=O)C1=CC(=C(C(=C1)O)O)O
Sulfated Ash 0.1% max.
Packaging Plastic bottle
Quantity 500g
Infrared Spectrum Authentic
Color White to Beige
Loss on Drying 0.5% max. (110°C, 4 hrs)
Melting Point 146.0°C to 150.0°C
Assay Percent Range 98%
Merck Index 15, 7971
Solubility Information Solubility in water: 0.35g/100 mL (25°C). Other solubilities: 103g/100 g alcohol,83g/100 g ether,soluble in aceton,propyleenglycol and,polysorbaat 8
Formula Weight 212.2
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Propyl gallate, 98%
Show More Show Less

GHS H Statement:
May cause an allergic skin reaction.
Harmful if swallowed.

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 204-498-2

RTECSNumber : LW8400000

TSCA : TSCA