Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propyl Hexanoate 98.0+%, TCI America™
SDP

Catalog No. P1940500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1940500ML 500 mL
P194025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1940500ML Supplier TCI America Supplier No. P1940500ML

Chemical Identifiers

CAS 626-77-7
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00053803
InChI Key HTUIWRWYYVBCFT-UHFFFAOYSA-N
Synonym propyl caproate, caproic acid propyl ester, hexanoic acid, propyl ester, n-propyl hexanoate, propionyl hexanoate, n-propyl n-hexanoate, hexanoic acid propyl ester, natural, fema no. 2949, n-propyl hexanoaten
PubChem CID 12293
ChEBI CHEBI:87365
IUPAC Name propyl hexanoate
SMILES CCCCCC(=O)OCCC

Specifications

Melting Point -69°C
Color Colorless
Boiling Point 187°C
Quantity 500 mL
UN Number 3272
Formula Weight 158.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl Hexanoate
EINECSNumber (2)-0782
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.