Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propyl propionate, 98+%

Catalog No. p-3593503
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle

CAS: 106-36-5 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-36-5
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009373
InChI Key MCSINKKTEDDPNK-UHFFFAOYSA-N
Synonym propyl propionate, n-propyl propionate, propanoic acid, propyl ester, n-propyl propanoate, propionic acid, propyl ester, n-propyl n-propionate, propylester kyseliny propionove, unii-g09trv00gk, propyl propionate natural, propionic acid n-propyl ester
PubChem CID 7803
IUPAC Name propyl propanoate
SMILES CCCOC(=O)CC

Specifications

Melting Point -76.0°C
Color Colorless
Density 0.8810g/mL
Boiling Point 123.0°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.3820 to 1.3940
Linear Formula CH3CH2C(O)O(CH2)2CH3
Quantity 50 mL
Specific Gravity 0.881
Merck Index 15, 7979
Solubility Information Solubility in water: soluble
Formula Weight 116.16
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Propyl propionate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Call a
EINECSNumber 203-389-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.