Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylamine hydrochloride, 98%

Catalog No. p-188492
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 556-53-6 | C3H10ClN | 95.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-53-6
Molecular Formula C3H10ClN
Molecular Weight (g/mol) 95.57
MDL Number MFCD00012911
InChI Key PYNUOAIJIQGACY-UHFFFAOYSA-N
Synonym propylamine hydrochloride, n-propylamine hydrochloride, 1-propanamine, hydrochloride, propylammonium chloride, propan-1-amine hydrochloride, 1-propylammonium chloride, n-propylammonium chloride, propylamine, hydrochloride, unii-a1z6ovy2hg, a1z6ovy2hg
PubChem CID 11165
IUPAC Name propan-1-amine;hydrochloride
SMILES [H+].[Cl-].CCCN

Specifications

Melting Point 160°C to 164°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3CH2CH2NH2·HCl
Quantity 25 g
Beilstein 04, 137
Merck Index 15, 7956
Formula Weight 95.57
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Propylamine hydrochloride, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-129-9
RTECSNumber UI3057000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.