Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. P194225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1942-25G 25 g
P1942-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1942-25G Supplier TCI America Supplier No. P194225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17745-45-8
Molecular Formula C3H9BO2
Molecular Weight (g/mol) 87.91
MDL Number MFCD01074564
InChI Key JAQOMSTTXPGKTN-UHFFFAOYSA-N
Synonym n-propylboronic acid, 1-propaneboronic acid, propaneboronic acid, boronic acid, propyl, 1-propylboronic acid
PubChem CID 351065
IUPAC Name propylboronic acid
SMILES CCCB(O)O

Specifications

Melting Point 106°C
Color White-Yellow
Quantity 25 g
Formula Weight 87.91
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.