Learn More
Propylene carbonate, 99%, Thermo Scientific Chemicals
Description
Propylene carbonate is a polar, aprotic solvent used in paints and coatings and as a high-permittivity component of electrolytes in lithium batteries. It plays an important role in adhesives, paint strippers, cosmetics, plasticizer, hard surface cleaner, resin cleaner, fiberglass, polyester and polyurethane cleaner. Furthermore, it acts as a cleaner in polyurethane, a carburetor and is used in stereo lithography parts. In electrospray ionization mass spectrometry, it is doped into low surface tension solutions to enhance analyte charging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Propylene carbonate is a polar, aprotic solvent used in paints and coatings and as a high-permittivity component of electrolytes in lithium batteries. It plays an important role in adhesives, paint strippers, cosmetics, plasticizer, hard surface cleaner, resin cleaner, fiberglass, polyester and polyurethane cleaner. Furthermore, it acts as a cleaner in polyurethane, a carburetor and is used in stereo lithography parts. In electrospray ionization mass spectrometry, it is doped into low surface tension solutions to enhance analyte charging.
Solubility
Miscible with water, alcohol, ether, acetone, benzene, chloroform and ethyl acetate.
Notes
Moisture sensitive. Hygroscopic. Incompatible with acids and bases.
Specifications
Melting Point | °C to 49°C |
Density | 1.204 |
Boiling Point | 240°C to 243°C |
Flash Point | 132°C (270°F) |
Assay Percent Range | 99% |
Refractive Index | 1.421 |
Quantity | 1000 g |
Solubility Information | Miscible with water,alcohol,ether,acetone,benzene,chloroform and ethyl acetate. |
Formula Weight | 102.09 |
Physical Form | Liquid |
Show More |
Safety and Handling
Hazard Category | H319 |
Precautionary Statement | P264b-P280i-P305+P351+P338 |
EINECSNumber | 203-572-1 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.