Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylene Carbonate, 99%, Spectrum™ Chemical
SDP

Catalog No. 18612968 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
500 g
3 kg
20 kg
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-968 20 kg Steel Can
18-612-966 25 g Amber Glass Bottle
18-612-970 500 g Amber Glass Bottle
18-612-969 200 L Drum
18-612-967 3 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-612-968 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P143320KG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 108-32-7

Specifications

CAS Max % 1
Packaging Steel Can
Quantity 20 kg
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.