Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylene Glycol (1,2-Propanediol), Reagents

Catalog No. 01337081
Click to view available options
Quantity:
1 L
4 L
20 L
200 L

Propylene Glycol (1,2-Propanediol), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 57-55-6
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.10
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
Synonym 1,2-Propanediol, Propane-1,2-diol
IUPAC Name propane-1,2-diol
SMILES CC(O)CO

Specifications

Boiling Range 188.2°C
Melting Point -60°C
Color Colorless
Boiling Point 188.2°C
Melting Range -60°C
Packaging Poly Drum
Quantity 200 L
Specific Gravity 1.036
Viscosity Viscous
Grade Reagent
Physical Form Liquid
Chemical Name or Material Propylene Glycol (1,2-Propanediol)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.