Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Propylene oxide, 99.5%, extra pure

Catalog No. p-3758726
Click to view available options
Quantity:
5 mL
1 L
2.5 L
Packaging:
Glass bottle

CAS: 75-56-9 | C3H6O | 58.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-56-9
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00005126
InChI Key GOOHAUXETOMSMM-UHFFFAOYNA-N
Synonym propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
PubChem CID 6378
ChEBI CHEBI:38685
IUPAC Name 2-methyloxirane
SMILES CC1CO1

Specifications

Melting Point -112.0°C
Density 0.8300g/mL
Boiling Point 34.0°C
Flash Point −37°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass bottle
Refractive Index 1.3650 to 1.3670
Quantity 5 mL
UN Number 1280
Beilstein 17, 6
Specific Gravity 0.83
Merck Index 15, 7969
Solubility Information Solubility in water: 40g/100mL (20°C). Other solubilities: miscible with alcohol and ether
Viscosity 0.32 mPa.s (20°C)
Formula Weight 58.08
Percent Purity 99.5%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material (±)-Propylene oxide, Extra Pure
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Extremely flammable liquid and vapor.
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
May cause cancer.
Precautionary Statement GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
EINECSNumber 200-879-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.