Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylenediphosphonic acid, 98+%

Catalog No. AA3027704
Click to view available options
Quantity:
0.5 g
2 g
10 g

CAS: 4671-82-3 | C3H10O6P2 | 204.055 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4671-82-3
Molecular Formula C3H10O6P2
Molecular Weight (g/mol) 204.055
MDL Number MFCD00050066
InChI Key DZQCAOQMXPROIJ-UHFFFAOYSA-N
Synonym propylenediphosphonic acid, acmc-20ajta, propylene-bisphosphonic acid, propylenediphosphonicacid, propane-1,2-diylbis phosphonic acid, 1-phosphonopropan-2-yl phosphonic acid
PubChem CID 22023450
IUPAC Name 1-phosphonopropan-2-ylphosphonic acid
SMILES CC(CP(=O)(O)O)P(=O)(O)O

Specifications

Melting Point 178°C
Odor Odorless
Quantity 2 g
Formula Weight 204.06
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Propylenediphosphonic acid
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.