Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylthiouracil, USP, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18605941
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail

P2075, 51-52-5, C7H10N2OS

Spectrum™ Chemical Propylthiouracil, USP is an anti-thyroid drug used to manage hyperthyroidism which is due to an overactive thyroid gland. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 51-52-5
Molecular Formula C7H10N2OS
Molecular Weight (g/mol) 170.23
MDL Number MFCD00006041
InChI Key KNAHARQHSZJURB-UHFFFAOYSA-N
IUPAC Name 6-propyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one
SMILES CCCC1=CC(=O)NC(=S)N1

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Melting Range 218-221°C
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98.0 to 100.5%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.