Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ProQuantum™ Immunoassays Accessory Kit

Catalog No. A36723
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A36723 10 assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A36723 Supplier Invitrogen™ Supplier No. A36723
Add to Cart
Add to Cart

The ProQuantum Immunoassay Accessory Kit provides the common consumable plastics to be used with any ProQuantum immunoassay product, conveniently bundled for first-time users who may be uncertain of what is needed or low-volume users who do not wish to buy these plastics in bulk.

Sufficient materials are provided to run 10 assays using 96-well plates. The kit does not include the following items: qPCR plates, cold blocks, and pipette tips.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Immunoassays Accessory Kit
Quantity 10 assays
Product Line ProQuantum
Content And Storage
  • 1 96-well plate rack
  • 10 25-mL reagent reservoirs for low volume pipetting
  • 20 adhesive seals
  • 10 optical seals
  • 1 hand seal applicator tool
  • 10 1.5-mL microfuge tubes
  • 10 15-mL conical tubes
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.