Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PROSAAS Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981583
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1583 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1583 Supplier Bioss Supplier No. BS11686R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene functions as an inhibitor of prohormone convertase 1, which regulates the proteolytic cleavage of neuroendocrine peptide precursors. The proprotein is further processed into multiple short peptides. A polymorphism within this gene may be associated with obesity.

Specifications

Antigen PROSAAS
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PCSK1N
Gene Accession No. Q9UHG2
Gene Alias AI848336; basic protein IA-4; Big LEN; big PEN LEN; Big PEN-LEN; Big SAAS; BigLEN; bLEN; b-LEN; BOS_25927; b-PEN-LEN; b-SAAS; granin-like neuroendocrine peptide; IA-4; KEP; Little LEN; Little SAAS; lLEN; l-LEN; l-SAAS; N-proSAAS; Pan3; pancreas protein 3; Pcsk1n; PEN; PEN19; PEN-19; PEN20; PEN-20; Proprotein convertase 1 inhibitor; Proprotein convertase subtilisin/kexin type 1 inhibitor; ProSAAS; pro-SAAS; protein serine alanine alanine serine; Saas; SAAS CT(1-49); SAAS CT(25-40); SAASCT; SCG8; SgVIII
Gene Symbols PCSK1N
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PROSAAS.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27344
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.