Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prosapip1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556614
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556614 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556614 Supplier Invitrogen™ Supplier No. PA556614
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QEFAMKSVGT RTGGGGSQGS FPGPRGSGSG ASRERPGRYP SEDKGLANSL YLNGELRGSD HTDVCGNVVG SSGGSSSSGG SDKAPPQYRE PS Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.

Prosapip1 is a binding partner of the postsynaptic density (PSD) protein ProSAP2/Shank3, a major scaffolding protein in excitatory brain synapses. Prosapip1 is thought to bind ProSAP2/Shank3 through a PDZ domain in ProSAP2/Shank3 and link it in turn to the Spine-associated Rap-Gap (SPAR). Prosapip1 shares significant homology with another PSD protein, PSD-Zip70, with both coding for a Fez1 domain that can be found in a total of four related protein, forming a novel family of scaffolding molecules termed 'Fezzins'. Recent studies show that Prosapip1 can be found in endocrine cells in which it is localized primarily in the hormone-storing compartment of these cells, suggesting that Prosapip1 may have other functions in non-neuronal cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prosapip1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Lzts3
Gene Accession No. O60299
Gene Alias KIAA0552; Leucine zipper putative tumor suppressor 3; leucine zipper tumor suppressor family member 3; leucine zipper, putative tumor suppressor family member 3; LZTS3; proline rich synapse associated protein interacting protein 1; proline rich synapse associated protein interacting protein 1 (Prosapip1); ProSAP/Shank-interacting protein 1; proSAP-interacting protein 1; PROSAPIP1
Gene Symbols Lzts3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LZTS3. Recombinant protein control fragment (Product #RP-95501).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9762
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.