Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ProSAPiP1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS11198R

Description
Prosapip1 is a binding partner of the postsynaptic density (PSD) protein ProSAP2/Shank3, a major scaffolding protein in excitatory brain synapses. Prosapip1 is thought to bind ProSAP2/Shank3 through a PDZ domain in ProSAP2/Shank3 and link it in turn to the Spine-associated Rap-Gap (SPAR). Prosapip1 shares significant homology with another PSD protein, PSD-Zip70, with both coding for a Fez1 domain that can be found in a total of four related protein, forming a novel family of scaffolding molecules termed ′Fezzins′. Recent studies show that Prosapip1 can be found in endocrine cells in which it is localized primarily in the hormone-storing compartment of these cells, suggesting that Prosapip1 may have other functions in non-neuronal cells.Specifications
ProSAPiP1 | |
Polyclonal | |
Unconjugated | |
Lzts3 | |
KIAA0552; Leucine zipper putative tumor suppressor 3; leucine zipper tumor suppressor family member 3; leucine zipper, putative tumor suppressor family member 3; LZTS3; proline rich synapse associated protein interacting protein 1; proline rich synapse associated protein interacting protein 1 (Prosapip1); ProSAP/Shank-interacting protein 1; ProSAP-interacting protein 1; PROSAPIP1 | |
Rabbit | |
Protein A | |
RUO | |
9762 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O60299 | |
Lzts3 | |
KLH conjugated synthetic peptide derived from human ProSAPiP1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction