Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prosapip2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA520814

Catalog No. PIPA520814


Only null left
Add to Cart

Description

Description

A suggested positive control is rat liver tissue lysate. PA5-20814 can be used with blocking peptide PEP-0928.

Prosapip2 (TBKBP1) is essential for signal transduction during viral infection thus plays a major role in the TNF/NF-kappa-B pathway. It is an adaptor protein that constitutively binds TBK1 (TANK-binding kinase) and IKBKE and may play a role in antiviral innate immunity. Prosapip2 is a 615 amino acid adaptor protein belonging to the fibrillar collagen family, consisting of trimers of identical alpha 1 chains which are linked to each other by interchain disulfide bonds. It has a ubiquitous expression with highest levels in ovary, followed by the neuronal system. Prosapip2 binds to TBK1 and helps in the activation of IRF3 which controls the expression of antiviral genes during infection. Recent studies show that Prosapip2 is an interaction partner of ProSAP2/Shank3 and actin, suggesting a role as a linker molecule between postsynaptic density and the cytoskeleton.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Prosapip2
Polyclonal
Unconjugated
TBKBP1
3110043L15Rik; KIAA0775; Prosapip2; Protein ProSAPiP2; RGD:631328}; Similar to NAP1 TBK1 adapter; Sintbad; TANK-binding kinase 1-binding protein 1; TBK1 binding protein 1; TBK1-binding protein 1; Tbkbp1; tbkbp1 {ECO:0000312
Rabbit
Antigen affinity chromatography
RUO
266764, 73174, 9755
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
A2A9T0, A7MCY6, Q6DG50
TBKBP1
A 16 amino acid peptide from near the carboxy terminus of human Prosapip2.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.