Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PROSC Antibody (OTI3G7), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273622APCCY7
Description
PROSC Monoclonal specifically detects PROSC in Human samples. It is validated for Flow Cytometry.Specifications
PROSC | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
PLPBP | |
Full length recombinant protein of human PROSC (NP_009129) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
OTI3G7 | |
Flow Cytometry | |
FLJ11861, proline synthase co-transcribed bacterial homolog protein, proline synthetase co-transcribed (bacterial homolog), proline synthetase co-transcribed bacterial homolog protein, proline synthetase co-transcribed homolog (bacterial) | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
11212.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction