Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PROSC Antibody (OTI3G7), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273622JF646
Description
PROSC Monoclonal specifically detects PROSC in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
| PROSC | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PLPBP | |
| Full length recombinant protein of human PROSC (NP_009129) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI3G7 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| FLJ11861, proline synthase co-transcribed bacterial homolog protein, proline synthetase co-transcribed (bacterial homolog), proline synthetase co-transcribed bacterial homolog protein, proline synthetase co-transcribed homolog (bacterial) | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 11212.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction