Learn More
Invitrogen™ PROSC Monoclonal Antibody (OTI1G2)

Description
PROSC Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
Specifications
| Antigen | PROSC |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1G2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PLPBP |
| Gene Accession No. | O94903 |
| Gene Alias | 1700024N20Rik; 2200002F22Rik; PLP homeostasis protein; PLPBP; Proline synthase co-transcribed bacterial homolog protein; proline synthetase co-transcribed; proline synthetase co-transcribed (bacterial homolog); proline synthetase co-transcribed bacterial homolog protein; proline synthetase co-transcribed homolog; proline synthetase cotranscribed homolog (bacterial); PROSC; Pyridoxal phosphate homeostasis protein; Pyridoxal phosphate-binding protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.