Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PROSC Monoclonal Antibody (OTI2C2), TrueMAB™, OriGene
SDP

Catalog No. 502539853
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9853 30 μL
50-167-5726 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9853 Supplier OriGene Supplier No. TA505195S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PROSC Monoclonal antibody specifically detects PROSC in Human samples. It is validated for Western Blot

Specifications

Antigen PROSC
Applications Western Blot
Classification Monoclonal
Clone OTI2C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PLPBP
Gene Accession No. O94903
Gene Alias 1700024N20Rik; 2200002F22Rik; PLP homeostasis protein; PLPBP; Proline synthase co-transcribed bacterial homolog protein; proline synthetase co-transcribed; proline synthetase co-transcribed (bacterial homolog); proline synthetase co-transcribed bacterial homolog protein; proline synthetase co-transcribed homolog; proline synthetase cotranscribed homolog (bacterial); PROSC; Pyridoxal phosphate homeostasis protein; Pyridoxal phosphate-binding protein
Host Species Mouse
Immunogen Full length human recombinant protein of human PROSC produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11212
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.