Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PROSC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532036
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532036 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532036 Supplier Invitrogen™ Supplier No. PA532036
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: THP-1. Predicted reactivity: Mouse (88%), Rat (88%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Pyridoxal 5'-phosphate (PLP)-binding protein, which may be involved in intracellular homeostatic regulation of pyridoxal 5'-phosphate (PLP), the active form of vitamin B6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PROSC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene PLPBP
Gene Accession No. O94903, Q9Z2Y8
Gene Alias 1700024N20Rik; 2200002F22Rik; PLP homeostasis protein; PLPBP; Proline synthase co-transcribed bacterial homolog protein; proline synthetase co-transcribed; proline synthetase co-transcribed (bacterial homolog); proline synthetase co-transcribed bacterial homolog protein; proline synthetase co-transcribed homolog; proline synthetase cotranscribed homolog (bacterial); PROSC; Pyridoxal phosphate homeostasis protein; Pyridoxal phosphate-binding protein
Gene Symbols PLPBP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PROSC. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11212, 114863
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.