Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PROSER1 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305784H
Description
PROSER1 Polyclonal antibody specifically detects PROSER1 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
PROSER1 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
C13orf23, Chromosome 13 Open Reading Frame 23, KIAA2032, Proline And Serine Rich 1, Proline And Serine-Rich Protein 1 | |
Produced in rabbits immunized with E. coli-derived Human PROSER1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunocytochemistry | |
HRP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
80209 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction