Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PROSER1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572025
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572025 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572025 Supplier Invitrogen™ Supplier No. PA572025
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PROSER1 Polyclonal Antibody for Western Blot

This gene encodes a conserved protein containing proline and serine rich regions. These regions may be important in protein-protein interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PROSER1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PROSER1
Gene Accession No. Q5PRE5, Q86XN7
Gene Alias 2810046L04Rik; 9330161F11; C13orf23; KIAA2032; proline and serine rich 1; proline and serine-rich protein 1; PROSER1
Gene Symbols PROSER1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 128-156 amino acids from the N-terminal region of human PROSER1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 212127, 80209
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.