Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ 50 Coveralls, Elastic Wrist and Ankles

Catalog No. 19065502H Shop All DuPont Personal Protection Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Height (English):
5 to 5.58 ft.
5.22 to 6.16 ft.
5.25 to 5.58 ft.
5.41 to 5.75 ft.
6 to 6.33 ft.
6.16 to 6.33 ft.
6.33 to 6.58 ft.
6.58 to 6.83 ft.
6.75 to 7.08 ft.
This item is not returnable. View return policy
Should not be used around heat, flame, sparks or potentially flammable or explosive environments.
This item is not returnable. View return policy
Should not be used around heat, flame, sparks or potentially flammable or explosive environments.

Microporous film coveralls provide protection from non-hazardous aerosols, light liquid splash, and dry particles.

  • Made with a microporous film that is laminated to a spundbonded polypropylene substrate
  • Front zipper closure
  • Elastic wrist
  • Elastic ankles
  • Serged seams
  • Collar
  • White
  • Thickness: 10 mil
TRUSTED_SUSTAINABILITY

Specifications

Product Type Single-use Coveralls
Size 5X-Large
Gender Unisex
Material Non-woven Fabric with Microporous Film
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Collar Type Laydown
Model NB125S
Size (English) Chest 54.75 to 58.25 in.
Size (English) Inseam 29.75 in.
Size (Metric) Inseam 75.56 cm
Thickness 10 mil
For Use With (Application) Janitorial, Sanitation, General Industrial, Maintenance
Length (English) Sleeve 40.5 in.
Length (Metric) Sleeve 102.87 cm
Width (English) Chest 34 in.
Width (Metric) Chest 86.36 cm
Height (English) 6.58 to 6.83 ft.
Height (Metric) 2.01 to 2.08 m
Quantity 25/Cs.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.