Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ PROSHIELD™ 6SFR Coverall
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
DuPont™ ProShield® 6 SFR garments are cost-efficient and designed to be worn over primary flame resistant clothing.
- Keeps dirt and grime off of primary flame resistant garments
- Provides a barrier against non-hazardous particles and aerosols

Specifications
Specifications
Size | 3X-Large |
Gender | Unisex |
Color | Blue |
Disposable | Single-use |
Ankle Style | Open |
Cuff Style | Open |
Neck Style | Collared |
Seam Type | Serged |
Sterility | Non-sterile |
Resistance | Flame, Particle |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction