Learn More
DuPont™ ProShield™ 80 Hooded Coverall
Catalog No. p-8893002 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
2X-Large
3X-Large
Large
Medium
Small
X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-986-613 Small
17-986-614 Medium
17-986-615 Large
17-986-616 X-Large
17-986-617 2X-Large
17-986-618 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-986-613 Supplier DuPont™ Supplier No. P8127BBUSM002500
This item is not returnable. View return policy
Provide protection against bloodborne pathogens.
  • Hydrophobic outer layer
  • Soft inner layer using bicomponent spunbond
  • Reinforced core layer as a redundant safety feature
  • Breathable fabric with MVTR measured according to ASTM D6701
  • Bound seams exceed the seam strength requirements defined in ASTM F3352
  • Comfort fit design with gusset crotch
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Coveralls
Size Small
Gender Unisex
Material ProShield
Color Blue
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Resistance Bloodborne Pathogens
Latex Free Yes
No. of Pockets None