Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ 10 Frock

Catalog No. 19162612 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Color:
Blue
White
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19-162-612 4X-Large Blue
19-162-614 Medium Blue
19-162-613 Large Blue
19-162-615 X-Large Blue
19-162-610 2X-Large Blue
19-162-611 3X-Large Blue
19-162-617 Medium White
19-162-616 Large White
19-162-618 X-Large White
19-162-653 2X-Large White
19-162-654 3X-Large White
19-162-655 4X-Large White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 19-162-612 Supplier DuPont™ Supplier No. PB271SBU4X003000
Add to Cart
Add to Cart
This item is not returnable. View return policy

Provides general protection against non-hazardous light liquid splashes and dry particulates.

  • Frock provides body-length jacket design with front zipper closure
  • Mandarin collar
  • Extends below hip
  • Elastic wrists
  • Serged seams
  • Manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY

Specifications

Size 4X-Large
Material ProShield
Color Blue
Closure Type Zipper
Cuff Style Elastic
No. of Pockets None
Length (Metric) 44.5
Seam Type Serged
For Use With (Application) General Protection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.