Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prostaglandin E Synthase Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299619B
Description
Prostaglandin E Synthase Polyclonal specifically detects Prostaglandin E Synthase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Prostaglandin E Synthase | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 5.3.99.3, MGC10317, MGST1L1MGST1-like 1, MGST1-L1mPGES-1, MGST-IV, Microsomal glutathione S-transferase 1-like 1, Microsomal prostaglandin E synthase 1, MPGES, MPGES1, MPGES-1, p53-induced apoptosis protein 12, p53-induced gene 12 protein, PGESmicrosomal prostaglandin E synthase-1, PIG12glutathione S-transferase 1-like 1, PP102, PP1294, prostaglandin E synthase, TP53I12, tumor protein p53 inducible protein 12 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Prostaglandin E Synthase. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Biotin | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
9536 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction