Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prostaglandin E1, 98+%

Catalog No. AC235020100
Change view
Click to view available options
Quantity:
10 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC235020100 10 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC235020100 Supplier Thermo Scientific Chemicals Supplier No. 235020100
Add to Cart
This item is not returnable. View return policy

CAS: 745-65-3 | C20H34O5 | 354.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 745-65-3
Molecular Formula C20H34O5
Molecular Weight (g/mol) 354.48
MDL Number MFCD00077860
InChI Key GMVPRGQOIOIIMI-DWKJAMRDSA-N
Synonym alprostadil, prostaglandin e1, pge1, muse, alprostadilum, prostavasin, topiglan, vasaprostan, femprox, edex
PubChem CID 5280723
ChEBI CHEBI:15544
IUPAC Name 7-[(1R,2R,3R)-3-hydroxy-2-[(E,3S)-3-hydroxyoct-1-enyl]-5-oxocyclopentyl]heptanoic acid
SMILES CCCCCC(C=CC1C(CC(=O)C1CCCCCCC(=O)O)O)O

Specifications

Melting Point 115.0°C to 116.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 10 mg
Merck Index 15, 7987
Specific Rotation Condition −64.00 (20.00°C c=1.0,C2H5OH)
Specific Rotation -64
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,methanol,ethyl acetate and,acetone
Formula Weight 354.48
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material Prostaglandin E1, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
Wear protective gloves/protective clothing/e
EINECSNumber 212-017-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.