Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prostaglandin E2, 98%

Catalog No. AC233560100
Change view
Click to view available options
Quantity:
10 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC233560100 10 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC233560100 Supplier Thermo Scientific Chemicals Supplier No. 233560100
Add to Cart
This item is not returnable. View return policy

CAS: 363-24-6 | C20H32O5 | 352.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 363-24-6
Molecular Formula C20H32O5
Molecular Weight (g/mol) 352.47
MDL Number MFCD00077861
InChI Key XEYBRNLFEZDVAW-ARSRFYASSA-N
Synonym prostaglandin e2, dinoprostone, pge2, prostin e2, prepidil, cervidil, propess, minprositin e2, minprostin e2, dinoproston
PubChem CID 5280360
ChEBI CHEBI:15551
IUPAC Name (Z)-7-[(1R,2R,3R)-3-hydroxy-2-[(E,3S)-3-hydroxyoct-1-enyl]-5-oxocyclopentyl]hept-5-enoic acid
SMILES CCCCCC(C=CC1C(CC(=O)C1CC=CCCCC(=O)O)O)O

Specifications

Melting Point 66°C to 68°C
Color Yellow to Amber
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 mg
Merck Index 15, 7988
Specific Rotation Condition −85.50 (20.00°C c=1,C2H5OH)
Specific Rotation -85.5
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in basic media,soluble in methanol,abs. ethanol,ethyl acetate,soluble in chloroform
Formula Weight 352.47
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material Prostaglandin E2, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-656-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.