CAS RN 363-24-6
Prostaglandin E2, 98%
CAS: 363-24-6 Molecular Formula: C20H32O5 Molecular Weight (g/mol): 352.47 MDL Number: MFCD00077861 InChI Key: XEYBRNLFEZDVAW-ARSRFYASSA-N Synonym: prostaglandin e2,dinoprostone,pge2,prostin e2,prepidil,cervidil,propess,minprositin e2,minprostin e2,dinoproston PubChem CID: 5280360 ChEBI: CHEBI:15551 IUPAC Name: (Z)-7-[(1R,2R,3R)-3-hydroxy-2-[(E,3S)-3-hydroxyoct-1-enyl]-5-oxocyclopentyl]hept-5-enoic acid SMILES: CCCCCC(C=CC1C(CC(=O)C1CC=CCCCC(=O)O)O)O
Prostaglandin E2 from Synthetic, MP Biomedicals™
CAS: 363-24-6 Molecular Formula: C20H32O5 Molecular Weight (g/mol): 352.47 MDL Number: MFCD00077861 InChI Key: XEYBRNLFEZDVAW-RTYMFESYNA-N Synonym: 5Z,11α,13E,15S]-11,15-Dihydroxy-9-oxoprosta-5,13-dien-1-oic acid,Dinoprostone,PGE₂
Prostaglandin E2, 99%, MP Biomedicals™
CAS: 363-24-6 Molecular Formula: C20H32O5 Molecular Weight (g/mol): 352.471 InChI Key: XEYBRNLFEZDVAW-ARSRFYASSA-N Synonym: prostaglandin e2,dinoprostone,pge2,prostin e2,prepidil,cervidil,propess,minprositin e2,minprostin e2,dinoproston PubChem CID: 5280360 ChEBI: CHEBI:15551 IUPAC Name: (Z)-7-[(1R,2R,3R)-3-hydroxy-2-[(E,3S)-3-hydroxyoct-1-enyl]-5-oxocyclopentyl]hept-5-enoic acid SMILES: CCCCCC(C=CC1C(CC(=O)C1CC=CCCCC(=O)O)O)O
MP Biomedicals™ Prostaglandin E2, γ-irradiated, ≥99%
Prostaglandin E2 is used in cell culture applications for the study of prostaglandin regulated cell signaling and gene regulation.
Prostaglandin E2 98.0+%, TCI America™
CAS: 363-24-6 Molecular Formula: C20H32O5 Molecular Weight (g/mol): 352.471 MDL Number: MFCD00077861 InChI Key: XEYBRNLFEZDVAW-ARSRFYASSA-N Synonym: prostaglandin e2,dinoprostone,pge2,prostin e2,prepidil,cervidil,propess,minprositin e2,minprostin e2,dinoproston PubChem CID: 5280360 ChEBI: CHEBI:15551 IUPAC Name: (Z)-7-[(1R,2R,3R)-3-hydroxy-2-[(E,3S)-3-hydroxyoct-1-enyl]-5-oxocyclopentyl]hept-5-enoic acid SMILES: CCCCCC(C=CC1C(CC(=O)C1CC=CCCCC(=O)O)O)O