Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Prostaglandin E2 Biotrak™ Assay

Catalog No. 45000887 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
96 Wells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-887 96 Wells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-887 Supplier Cytiva Supplier No. RPN222

Includes

RPN222 supplied with sufficient quantities of reagents for 96 wells; RPN22210, for 960 wells. RPN22210 does not include lysis reagents. Both allow for construction of one standard curve and measurement of 37 unknowns in duplicate per plate.

Enzyme immunoassay system suitable for plasma, urine, or cell culture supernatant samples

  • Specific for PGE2
  • Flexible method with choice of five assay protocols
  • Product available with or without lysis reagents
  • Lysis reagents facilitate extraction of PGEsubscript 2 from cell cultures and plasma samples; PGE is directly available for subsequent analysis without removal of extraction reagents
  • Precise and accurate measurement
  • Ready to use substrate and color-coded reagents
  • Range: 20-640pg/mL (1-32pg/well)
  • Room temperature protocol: 50-6400pg/mL
  • Sensitivity: 16pg/mL, 0.8pg/well
  • 5-hour protocol
  • Storage: Store at -15° to -30°C.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 20 to 640pg/mL
Includes Microplate (12 x 8-well strip coated with sheep Anti-Mouse IgG), Assay Buffer, standard (prostaglandin E2) 256ng/mL, prostaglandin E Antibody (lyophilized), Prostaglandin E2 Conjugate, Wash Buffer, TMB Substrate, lysis reagent 1, lysis reagent 2
Quantity 96 Wells
Sample Type Plasma (EDTA, Citrate), Urine, Tissue Homogenates, Cell Culture, Cell Culture Supernatant
Sample Volume 50 μL
Sensitivity 16 pg/mL (0.8 pg/Well)
Total Assay Time 5 hr.
Content And Storage -15°C to -30°C
For Use With (Application) ELISA
Product Type Prostaglandin E2 Direct Biotrak assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.