Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prostaglandin E2 from Synthetic, MP Biomedicals™

Catalog No. MP219546105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219546105 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219546105 Supplier MP Biomedicals, Inc Supplier No. 0219546105
Add to Cart
Add to Cart

Prostaglandin E2 from Synthetic ≥99%, White Powder

  • Prostaglandin E2 induces cervical ripening and parturition;
  • mediates bradykinin-induced vasodilation.
  • PGE2 is one of the primary cyclooxygenase products of arachidonic acid and one of the most widely investigated prostaglandins.
  • Its activity influences inflammation, fertility and parturition, gastric mucosal integrity, and immune modulation.
  • The effects of PGE2 are transduced by at least four distinct receptors designated EP1, EP2, EP3, and EP4.
  • Affinity constants (Kd) of PGE2 for these receptors range from 1-10 nM depending on the receptor subtype and tissue.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 363-24-6
Molecular Formula C20H32O5
Molecular Weight (g/mol) 352.47
MDL Number MFCD00077861
InChI Key XEYBRNLFEZDVAW-RTYMFESYNA-N
Synonym 5Z,11α,13E,15S]-11,15-Dihydroxy-9-oxoprosta-5,13-dien-1-oic acid, Dinoprostone, PGE₂

Specifications

Melting Point 66°C to 68°C (literature)
Color White
Quantity 5 mg
Beilstein 4709356
Solubility Information Sparingly soluble in water but freely soluble in organic solvents such as ethanol, methanol, acetone (clear solution), acetonitrile, or DMSO. The solubility of PGE2 in these solvents is at least 10mg/mL
Physical Form Powder
Grade ≥99%
RTECSNumber UK8000000
storageNote1 Sparingly soluble in water but freely soluble in organic solvents such as ethanol, methanol, acetone (clear solution), acetonitrile, or DMSO. The solubility of PGE2 in these solvents is at least 10mg/mL.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.