Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prostaglandin E2 98.0+%, TCI America™
SDP

Catalog No. P188410MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P188410MG 10 mg
P18841MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P188410MG Supplier TCI America Supplier No. P188410MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 363-24-6
Molecular Formula C20H32O5
Molecular Weight (g/mol) 352.471
MDL Number MFCD00077861
InChI Key XEYBRNLFEZDVAW-ARSRFYASSA-N
Synonym prostaglandin e2, dinoprostone, pge2, prostin e2, prepidil, cervidil, propess, minprositin e2, minprostin e2, dinoproston
PubChem CID 5280360
ChEBI CHEBI:15551
IUPAC Name (Z)-7-[(1R,2R,3R)-3-hydroxy-2-[(E,3S)-3-hydroxyoct-1-enyl]-5-oxocyclopentyl]hept-5-enoic acid
SMILES CCCCCC(C=CC1C(CC(=O)C1CC=CCCCC(=O)O)O)O

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 10 mg
Formula Weight 352.47
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Prostaglandin E2
RTECSNumber UK8000000
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.