Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Prostaglandin F2 alpha ELISA Kit (Colorimetric)
SDP

Catalog No. NBP262167
Click to view available options
Quantity:
1 Kit
5 Kit

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Non-species specific Prostaglandin F2 alpha ELISA Kit (Colorimetric) measures non-species specific Prostaglandin F2 alpha in culture supernatants, plasma, serum, saliva, urine, cell lysate, whole blood.

1 Kit

Specifications

Assay Range 3.05 - 50,000 pg/ml
Assay Sensitivity 6.71 pg/ml
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Lysates, Cell Culture Supernatant, Plasma, Saliva, Serum, Urine, Whole Blood
For Use With (Application) ELISA
Host Species Non-species specific
Quantity 1 Kit
Storage Requirements Store at 4°C
Target Prostaglandin F2 alpha
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.