Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Prostaglandin F2alpha
GREENER_CHOICE

Catalog No. 421410
Change view
Click to view available options
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
42-141-0 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 42-141-0 Supplier Tocris Bioscience™ Supplier No. 4214/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Naturally-occurring prostanoid; potent vasoconstrictor

Prostaglandin F2α is a prostaglandin E2 and F2α receptor agonist. Naturally-occurring prostaglandin and potent vasoconstrictor. Induces cardiomyocyte hypertrophy in vitro. Exhibits luteolytic activity.

Specifications

Quantity 10 mg
Product Type Prostaglandin F
Purity >99%
Molecular Weight (g/mol) 475.62
Inhibitors Prostaglandin F

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.