Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prostaglandin I2 Receptor Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981253
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1253 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1253 Supplier Bioss Supplier No. BS11307R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Receptor for prostacyclin (prostaglandin I2 or PGI2). The activity of this receptor is mediated by G(s) proteins which activate adenylate cyclase.

Specifications

Antigen Prostaglandin I2 Receptor
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PTGIR
Gene Accession No. P43119
Gene Alias IP; IP receptor; PGI receptor; PGI2; PGI2 receptor; PRIPR; Prostacyclin receptor; prostaglandin I receptor (IP); prostaglandin I2 (prostacyclin) receptor (IP); Prostaglandin I2 receptor; Prostanoid IP receptor; Ptgir
Gene Symbols PTGIR
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Prostaglandin I2 Receptor.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5739
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.