Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prostasin/Prss8 Antibody (107), Alexa Fluor™ 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289382AF488
Description
Prostasin/Prss8 Monoclonal antibody specifically detects Prostasin/Prss8 in Human samples. It is validated for ELISASpecifications
Prostasin/Prss8 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
5652 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
107 | |
ELISA | |
CAP1, channel-activating protease 1, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.120, PROSTASIN, protease, serine, 8, Serine protease 8 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Prostasin/Prss8 (NP_002764.1; Ala 30-Arg 322). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction