Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prostate and colon associated protein Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987463
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7463 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7463 Supplier Bioss Supplier No. BS6742R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It belongs to the TPD52 family. Its exact function remains unknown.

Specifications

Antigen Prostate and colon associated protein
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Tpd52
Gene Accession No. P55327
Gene Alias 28 kDa calcium-dependent phosphoprotein; AI043038; BOS_13930; calcium sensitive phosphoprotein (CSPP28); CSPP28; D52; hD52; mD52; N8L; PC-1; pp28; PrLZ; prostate and colon associated protein; prostate leucine zipper; Protein N8; si:ch211-203l9.2; si:ch211-203l9.3; Tpd52; tumor promoter D52 protein; Tumor protein D52
Gene Symbols Tpd52
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TPD52.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7163
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.