Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prostate Specific Acid Phosphatase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113743
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113743 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113743 Supplier Invitrogen™ Supplier No. PA5113743
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CESVHNFTLP SWATEDTMTK LRELSELSLL SLYGIHKQKE KSRLQGGVLV For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

ACPP is an enzyme which catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate. It is synthesized under androgen regulation and is secreted by the epithelial cells of the prostate gland.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prostate Specific Acid Phosphatase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ACP3
Gene Accession No. P15309
Gene Alias 5'-NT; 5'-nucleotidase; A030005E02Rik; acid phosphatase, prostate; ACP3; ACP-3; Acpp; Acpp11; EC 3.1.3.2; Ecto-5'-nucleotidase; fluoride-resistant acid phosphatase; FRAP; Lap; lysosomal acid phosphatase; PAP; PAPf39; Ppal; PPAP; prostate acid phosphatase; Prostatic acid phosphatase; prostatic acid phosphatase (rPAP); prostatic acid phosphotase; RNACPP11; Thiamine monophosphatase; TMPase
Gene Symbols ACP3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ACPP.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.