Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Prostate Specific Acid Phosphatase (PSAP) Ab-1, Mouse Monoclonal Antibody

Catalog No. MS321P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS321P1 500 μL Concentrated 200μg/mL
MS321P1ABX 100 μL Concentrated 1mg/mL
MS321PABX 200 μL Concentrated 1mg/mL
MS321P 1 mL Concentrated 200μg/mL
MS321R1 1 mL Prediluted -
MS321RQ 12 mL Ready-to-Use -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS321P1 Supplier Epredia™ Supplier No. MS321P1

Provide accurate, reproducible results with the Epredia™ Prostate Specific Acid Phosphatase (PSAP) Ab-1, Mouse Monoclonal Antibody.

Prostatic acid phosphatase (PSAP) is one of the two antigenic markers of prostatic carcinoma, the other being prostate specific antigen. It belongs to the kallikrein family of serine proteases and is suggested to act as a hydrolase to split phospharyl choline in semen and as a transferase.

Host Species: Mouse

Clone: PASE/4LJ

Isotype: IgG1

Species Reactivity: Human. Does not react with dog, rabbit, and rat. Others not known.

Epitope: Not determined

Immunogen: Purified prostatic acid phosphatase (PSAP) from human seminal plasma

Molecular Weight: 52kDa

Positive Control: Normal prostate or prostate CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Specifications

Antigen Prostate Specific Acid Phosphatase (PSAP) Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PASE/4LJ
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified prostatic acid phosphatase (PSAP) from human seminal plasma
Quantity 500 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.