Learn More
Epredia™ Lab Vision™ Prostate Specific Acid Phosphatase (PSAP) Ab-1, Mouse Monoclonal Antibody
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: PASE/4LJ
Isotype: IgG1
Species Reactivity: Human. Does not react with dog, rabbit, and rat. Others not known.
Epitope: Not determined
Immunogen: Purified prostatic acid phosphatase (PSAP) from human seminal plasma
Molecular Weight: 52kDa
Positive Control: Normal prostate or prostate CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Specifications
Specifications
| Antigen | Prostate Specific Acid Phosphatase (PSAP) Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | PASE/4LJ |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Purified prostatic acid phosphatase (PSAP) from human seminal plasma |
| Quantity | 12 mL |
| Regulatory Status | IVD |
| Research Discipline | Cancer and Tumor Biology |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.