Learn More
Invitrogen™ Prostate Specific Acid Phosphatase Recombinant Rabbit Monoclonal Antibody (202)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA530059
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse Prostatic Acid Phosphatase/ACPP.
ACPP is an enzyme which catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate. It is synthesized under androgen regulation and is secreted by the epithelial cells of the prostate gland.
Specifications
Prostate Specific Acid Phosphatase | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q8CE08 | |
ACP3 | |
Recombinant Mouse Prostatic Acid Phosphatase/ACPP protein (Met1-Arg381). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
ELISA | |
202 | |
Unconjugated | |
ACP3 | |
5'-NT; 5'-nucleotidase; A030005E02Rik; acid phosphatase, prostate; ACP3; ACP-3; Acpp; Acpp11; EC 3.1.3.2; Ecto-5'-nucleotidase; fluoride-resistant acid phosphatase; FRAP; Lap; lysosomal acid phosphatase; PAP; PAPf39; Ppal; PPAP; prostate acid phosphatase; Prostatic acid phosphatase; prostatic acid phosphatase (rPAP); prostatic acid phosphotase; RNACPP11; Thiamine monophosphatase; TMPase | |
Rabbit | |
Protein A | |
RUO | |
56318 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.