Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Prostate Specific Antigen (PSA), Rabbit Monoclonal Antibody

Catalog No. RM2104S0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM2104S0 100 μL Concentrated
RM2104S 1 mL Concentrated
RM2104R7 7 mL Prediluted
RM2104R1 1 mL Prediluted
RM2104RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RM2104S0 Supplier Epredia™ Supplier No. RM2104S0

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Employ part of a comprehensive antibody portfolio Epredia™ Prostate Specific Antigen (PSA), Rabbit Monoclonal Antibody. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

PSA is a chymotrypsin-like serine protease (kallikrein family) produced by the prostate epithelium, and abundant in seminal fluid. PSA can be detected in the sera of patients with prostatic carcinoma. It is predominantly complexed to a liver-derived serine protease inhibitor, alpha-1-antichymotrypsin (ACT). A higher proportion of serum PSA is complexed to ACT in prostate cancer than in benign prostate hyperplasia. PSA is used to confirm prostatic acinar cell origin in primary and metastatic carcinoma and to rule out non-prostatic carcinoma mimics.

Host Species: Rabbit

Clone: EP1588Y

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide corresponding to residues near the C-terminus of human PSA

Molecular Weight: 34kDa

Positive Control: Normal prostate or prostate carcinoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Prostate Specific Antigen (PSA)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EP1588Y
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues near the C-terminus of human PSA
Quantity 100 μL
Regulatory Status ASR
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.