Learn More
Description
Specifications
Specifications
| Quantity | 5 Slides |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Tissue Condition | Normal |
| Cross Reactivity | Fas Receptor/TNFRSF6/CD95 |
| Host Species | Human |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Life Stage | Adult |
| Product Line | Novus |
| Tissue | Brain |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
