Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prostate Tumor Monoclonal Antibody (YPMA-2)
GREENER_CHOICE

Catalog No. AMPB1390
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AMPB1390 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AMPB1390 Supplier Invitrogen™ Supplier No. MA121390
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts specifically with prostate mucin antigen. It does not react with fetal or benign prostate specimens, non-prostate carcinomas and normal human tissues.

Prostate tumors, primarily arising as prostate cancer, are one of the most common malignancies in men. Prostate cancer develops in the prostate gland, which is responsible for producing seminal fluid. The onset of prostate cancer is often asymptomatic in its early stages, but as the disease progresses, symptoms such as difficulty in urination, pelvic pain, and blood in urine or semen may appear. Prostate cancer is typically a slow-growing tumor; however, some forms can be aggressive and metastasize to other parts of the body, particularly the bones and lymph nodes. The development of prostate cancer is influenced by genetic, environmental, and hormonal factors, with age being a significant risk factor. Diagnosis is commonly achieved through digital rectal exams (DRE), prostate-specific antigen (PSA) testing, and biopsy. Treatment options vary depending on the stage and aggressiveness of the tumor, ranging from active surveillance for low-risk cases to surgery, radiation therapy, hormonal therapy, and chemotherapy for more advanced stages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prostate Tumor
Applications Western Blot
Classification Monoclonal
Clone YPMA-2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.2
Gene Alias pma; Prostate Mucin Antigen
Host Species Mouse
Immunogen Membrane protein extracted from human prostate mucin tissues.
Purification Method Protein G
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.