Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prostatic Acid Phosphatase/ACPP Antibody (205), Alexa Fluor™ 647

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290773AF647
Description
Prostatic Acid Phosphatase/ACPP Monoclonal specifically detects Prostatic Acid Phosphatase/ACPP in Mouse samples. It is validated for Sandwich ELISA Detection.Specifications
Prostatic Acid Phosphatase/ACPP | |
Monoclonal | |
Alexa Fluor 647 | |
acid phosphatase, prostate, ACP3, ACP-3, EC 3.1.3.2, PAP, prostatic acid phosphatase, prostatic acid phosphotase | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Prostatic Acid Phosphatase/ACPP (Q8CE08-1; Met1-Arg381). | |
0.1 mL | |
Cell Cycle and Replication | |
55 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
205 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction