Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prostatic Acid Phosphatase/ACPP Antibody (SPM312), Novus Biologicals™
SDP

Catalog No. NB253169100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253169100 100 μg
NB25316920 20 μg
NB253169200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB253169100 Supplier Novus Biologicals Supplier No. NBP253169100UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Prostatic Acid Phosphatase/ACPP Monoclonal specifically detects Prostatic Acid Phosphatase/ACPP in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Prostatic Acid Phosphatase/ACPP
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM312
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Gene Alias acid phosphatase, prostate, ACP3, ACP-3, EC 3.1.3.2, PAP, prostatic acid phosphatase, prostatic acid phosphotase
Gene Symbols ACPP
Host Species Mouse
Immunogen Prostatic acid phosphatase purified from human seminal plasma
Molecular Weight of Antigen 52 kDa
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 55
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.