Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prostatic Acid Phosphatase/ACPP Rabbit anti-Mouse, Clone: 202, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $524.00
Specifications
Antigen | Prostatic Acid Phosphatase/ACPP |
---|---|
Clone | 202 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
Prostatic Acid Phosphatase/ACPP Monoclonal specifically detects Prostatic Acid Phosphatase/ACPP in Mouse samples. It is validated for ELISA.Specifications
Prostatic Acid Phosphatase/ACPP | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
acid phosphatase, prostate, ACP3, ACP-3, EC 3.1.3.2, PAP, prostatic acid phosphatase, prostatic acid phosphotase | |
Recombinant Mouse Prostatic Acid Phosphatase/ACPP protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
202 | |
ELISA | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
0.2 um filtered solution in PBS | |
55 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title